Assay Details
Target Gene Details
Entrez Gene ID: | 11101 |
Gene Name: | arginyltransferase 1 |
Gene Aliases: |
- |
Location: |
Chr.10:121740421-121928801 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATE1 | NM_001001976.2 | 12 | 2157 | NP_001001976.1 |
| NM_001288734.1 | 11 | 2053 | NP_001275663.1 | |
| NM_001288735.1 | 10 | 2030 | NP_001275664.1 | |
| NM_001288736.1 | 12 | 2392 | NP_001275665.1 | |
| NM_007041.3 | 12 | 2157 | NP_008972.2 | |
| NR_110006.1 | 10 | 2099 | ||
| XM_005269458.2 | 13 | 2298 | XP_005269515.1 | |
| XM_005269459.3 | 13 | 2521 | XP_005269516.1 | |
| XM_017015597.1 | 10 | 1967 | XP_016871086.1 | |
| XM_017015598.1 | 12 | 2376 | XP_016871087.1 | |
| XM_017015599.1 | 11 | 2040 | XP_016871088.1 | |
| XM_017015600.1 | 12 | 2020 | XP_016871089.1 | |
| XM_017015602.1 | 11 | 1937 | XP_016871091.1 | |
| XM_017015604.1 | 10 | 1882 | XP_016871093.1 | |
| AF079098.1 | AAD12366.1 | |||
| AF079099.1 | AAD12367.1 | |||
| AK124669.1 | 10 | 2107 | ||
| AK295642.1 | ||||
| AK303604.1 | ||||
| BC022026.1 | 12 | 2130 | AAH22026.2 |
More Information
Set Membership: |
|

Genomic Map