Assay Details
Target Gene Details
Entrez Gene ID: | 146850 |
Gene Name: | phosphoinositide-3-kinase regulatory subunit 6 |
Gene Aliases: |
C17orf38, HsT41028, p84 PIKAP, p87(PIKAP), p87PIKAP |
Location: |
Chr.17:8802723-8870003 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3R6 | NM_001010855.3 | 20 | 2411 | NP_001010855.1 |
| NM_001290211.1 | 20 | 2779 | NP_001277140.1 | |
| NR_110865.1 | 20 | 2647 | ||
| XM_011523677.2 | 20 | 2305 | XP_011521979.1 | |
| XM_011523678.2 | 20 | 2292 | XP_011521980.1 | |
| XM_011523680.2 | 19 | 2185 | XP_011521982.1 | |
| XM_011523681.2 | 19 | 2556 | XP_011521983.1 | |
| XM_011523683.2 | 17 | 1993 | XP_011521985.1 | |
| XM_011523686.2 | 11 | 1442 | XP_011521988.1 | |
| XM_017024228.1 | 20 | 2307 | XP_016879717.1 | |
| XM_017024229.1 | 21 | 2423 | XP_016879718.1 | |
| XM_017024230.1 | 20 | 2300 | XP_016879719.1 | |
| XM_017024231.1 | 16 | 2484 | XP_016879720.1 | |
| AK091819.1 | 20 | 2649 | ||
| AL833037.1 | 1 | 148 | CAH56340.1 | |
| AY753192.1 | 18 | 2171 | AAV30085.1 | |
| BC125169.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521208 | Chr.17:8803133 - 8803711 on Build GRCh38 | Loss |
|
| esv3639892 | Chr.17:8800713 - 8805464 on Build GRCh38 | Loss |
|
| nsv518675 | Chr.17:8696386 - 8818290 on Build GRCh38 | Gain |
|
| nsv953828 | Chr.17:8796283 - 8806582 on Build GRCh38 | Deletion |
|
| nsv1969 | Chr.17:8780681 - 8825867 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map