Assay Details
Target Gene Details
Entrez Gene ID: | 113201 |
Gene Name: | cancer susceptibility candidate 4 |
Gene Aliases: |
H63 |
Location: |
Chr.15:44288616-44415761 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASC4 | NM_138423.3 | NP_612432.2 | ||
| NM_177974.2 | NP_816929.1 | |||
| XM_017021880.1 | XP_016877369.1 | |||
| AK098265.1 | BAC05272.1 | |||
| AK291026.1 | ||||
| AK298564.1 | ||||
| AL832333.1 | ||||
| AL832840.1 | ||||
| AY358086.1 | AAQ88453.1 | |||
| BC012124.1 | AAH12124.1 | |||
| BC063480.1 | AAH63480.1 | |||
| DB229269.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2587n100 | Chr.15:44197336 - 44405284 on Build GRCh38 | Gain |
|

Genomic Map