Assay Details
Target Gene Details
Entrez Gene ID: | 56922 |
Gene Name: | methylcrotonoyl-CoA carboxylase 1 |
Gene Aliases: |
MCC-B, MCCA |
Location: |
Chr.3:183015218-183099587 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCCC1 | NM_001293273.1 | NP_001280202.1 | ||
| NM_020166.4 | NP_064551.3 | |||
| NR_120639.1 | ||||
| NR_120640.1 | ||||
| XM_006713702.1 | XP_006713765.1 | |||
| XM_011512992.2 | XP_011511294.1 | |||
| XM_017006874.1 | XP_016862363.1 | |||
| XM_017006875.1 | XP_016862364.1 | |||
| AB029826.1 | BAA99407.1 | |||
| AB209737.1 | BAD92974.1 | |||
| AF297332.1 | AAK67986.1 | |||
| AF310339.1 | AAG50245.1 | |||
| AF310972.1 | AAG53095.1 | |||
| AK023051.1 | BAB14377.1 | |||
| AK098411.1 | ||||
| AK294882.1 | ||||
| AL442091.1 | ||||
| BC004187.2 | AAH04187.1 | |||
| BC004214.2 | AAH04214.1 | |||
| CR749608.1 | CAH18403.1 | |||
| KU178727.1 | ||||
| KU178728.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv592654 | Chr.3:182938822 - 183025479 on Build GRCh38 | Loss |
|
| nsv482968 | Chr.3:182869307 - 183035390 on Build GRCh38 | Gain |
|
| nsv508968 | Chr.3:182974407 - 183063558 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map