Assay Details
Target Gene Details
Entrez Gene ID: | 7091 |
Gene Name: | transducin like enhancer of split 4 |
Gene Aliases: |
BCE-1, BCE1, E(spI), ESG, ESG4, GRG4, Grg-4 |
Location: |
Chr.9:79571773-79726882 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831634 | Chr.9:79430279 - 79589786 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map