Assay Details
Target Gene Details
Entrez Gene ID: | 58477 |
Gene Name: | SRP receptor beta subunit |
Gene Aliases: |
APMCF1 |
Location: |
Chr.3:133784033-133821492 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRPRB | NM_021203.3 | 8 | 1081 | NP_067026.3 |
| AF086914.1 | 7 | 819 | AAP97144.1 | |
| AF141882.3 | 7 | 812 | AAD34888.3 | |
| AK027307.1 | 4 | 1324 | ||
| AK027525.1 | 7 | 812 | BAB55176.1 | |
| AK075531.1 | 7 | 850 | BAC11675.1 | |
| BC063001.1 | 7 | 824 | AAH63001.1 | |
| BC065299.1 | 7 | 796 | AAH65299.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829729 | Chr.3:133726027 - 133901543 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map