Assay Details
Target Gene Details
Entrez Gene ID: | 2274 |
Gene Name: | four and a half LIM domains 2 |
Gene Aliases: |
AAG11, DRAL, FHL-2, SLIM-3, SLIM3 |
Location: |
Chr.2:105360826-105438773 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
More Information
Set Membership: |
|

Genomic Map