Assay Details
Target Gene Details
Entrez Gene ID: | 311 |
Gene Name: | annexin A11 |
Gene Aliases: |
ANX11, CAP50 |
Location: |
Chr.10:80153953-80205677 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551635 | Chr.10:79925268 - 80212630 on Build GRCh38 | Loss |
|
| esv3623946 | Chr.10:80133465 - 80175734 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map