Assay Details
Target Gene Details
Entrez Gene ID: | 50636 |
Gene Name: | anoctamin 7 |
Gene Aliases: |
D-TMPP, DTMPP, IPCA-5, IPCA5, NGEP, PCANAP5, PCANAP5L, TMEM16G |
Location: |
Chr.2:241188509-241239602 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANO7 | NM_001001666.3 | NP_001001666.1 | ||
| NM_001001891.3 | NP_001001891.2 | |||
| XM_011511263.2 | XP_011509565.1 | |||
| XM_011511267.2 | XP_011509569.1 | |||
| XM_017004229.1 | XP_016859718.1 | |||
| AY617079.1 | AAT40139.1 | |||
| AY617080.1 | AAT40140.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3241 | Chr.2:241158269 - 241190667 on Build GRCh38 | Insertion |
|
| nsv527790 | Chr.2:241184874 - 241223944 on Build GRCh38 | Loss |
|
| nsv508900 | Chr.2:241177974 - 241244786 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map