Assay Details
Target Gene Details
Entrez Gene ID: | 23603 |
Gene Name: | coronin 1C |
Gene Aliases: |
HCRNN4 |
Location: |
Chr.12:108645109-108731550 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CORO1C | NM_001105237.2 | 11 | 3667 | NP_001098707.1 |
| NM_001276471.1 | 11 | 3685 | NP_001263400.1 | |
| NM_014325.3 | 11 | 3586 | NP_055140.1 | |
| XM_011538125.1 | 7 | 2910 | XP_011536427.1 | |
| XM_017019121.1 | 12 | 4415 | XP_016874610.1 | |
| XM_017019122.1 | 11 | 3992 | XP_016874611.1 | |
| XM_017019123.1 | 12 | 4045 | XP_016874612.1 | |
| AL162070.1 | 11 | 3524 | CAB82406.1 | |
| BM677390.1 | 1 | 273 | ||
| Z24726.1 | 1 | 726 |
More Information
Set Membership: |
|

Genomic Map