Assay Details
Target Gene Details
Entrez Gene ID: | 22879 |
Gene Name: | MON1 homolog B, secretory trafficking associated |
Gene Aliases: |
HSRG1, SAND2, SRG1 |
Location: |
Chr.16:77190919-77199646 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MON1B | NM_001286639.1 | 3 | 881 | NP_001273568.1 |
| NM_001286640.1 | 2 | 723 | NP_001273569.1 | |
| NM_014940.3 | 4 | 1208 | NP_055755.1 | |
| AB020679.1 | 4 | 892 | ||
| AF442486.1 | 3 | 872 | AAL35292.1 | |
| AK023374.1 | ||||
| AK023896.1 | 4 | 936 | ||
| AK128411.1 | 1 | 750 | BAC87427.1 | |
| AK293392.1 | 2 | 443 | ||
| AK296467.1 | 3 | 608 | ||
| AW503582.1 | 1 | 58 | ||
| BC024277.1 | 4 | 1188 | AAH24277.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056676 | Chr.16:77105785 - 77820951 on Build GRCh38 | Loss |
|
| nsv1055234 | Chr.16:76969529 - 77433389 on Build GRCh38 | Gain |
|
| dgv3027n100 | Chr.16:76614676 - 77688151 on Build GRCh38 | Gain |
|
| nsv523187 | Chr.16:77193283 - 77195760 on Build GRCh38 | Loss |
|
| nsv1115510 | Chr.16:77192403 - 77196203 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map