Assay Details
Target Gene Details
Entrez Gene ID: | 623 |
Gene Name: | bradykinin receptor B1 |
Gene Aliases: |
B1BKR, B1R, BDKRB2, BKB1R, BKR1, BRADYB1 |
Location: |
Chr.14:96256210-96264763 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BDKRB1 | NM_000710.3 | 3 | 748 | NP_000701.2 |
| AJ238044.1 | 3 | 731 | CAB45650.1 | |
| AK291970.1 | 3 | 749 | ||
| AK309528.1 | 3 | 748 | ||
| BC034705.1 | 2 | 654 | AAH34705.1 | |
| KJ950627.1 | 4 | 882 | ||
| KJ950628.1 | 3 | 753 | ||
| U12512.1 | 1 | 534 | AAA21346.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv565648 | Chr.14:96263509 - 96266550 on Build GRCh38 | Loss |
|
| esv3635420 | Chr.14:96224959 - 96319922 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map