Assay Details
Target Gene Details
Entrez Gene ID: | 64175 |
Gene Name: | prolyl 3-hydroxylase 1 |
Gene Aliases: |
GROS1, LEPRE1, OI8 |
Location: |
Chr.1:42746335-42767084 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P3H1 | NM_001146289.1 | 15 | 2344 | NP_001139761.1 |
| NM_001243246.1 | 14 | 2744 | NP_001230175.1 | |
| NM_022356.3 | 15 | 2325 | NP_071751.3 | |
| XM_005271110.2 | 14 | 1662 | XP_005271167.1 | |
| XM_011541948.1 | 15 | 1717 | XP_011540250.1 | |
| XM_017002051.1 | 16 | 2297 | XP_016857540.1 | |
| XM_017002052.1 | 16 | 2294 | XP_016857541.1 | |
| AF097431.1 | 15 | 2492 | AAG31018.1 | |
| AF097432.1 | 15 | 2263 | AAG31019.1 | |
| AK025841.1 | 11 | 1181 | BAB15256.1 | |
| AK027648.1 | 14 | 2672 | BAB55264.1 | |
| AK027680.1 | 15 | 2227 | BAB55291.1 | |
| AK027697.1 | 15 | 2264 | BAB55305.1 | |
| AK075418.1 | 15 | 2242 | BAC11608.1 | |
| AK297980.1 | 16 | 2130 | ||
| BC015309.2 | 11 | 1227 | AAH15309.2 | |
| BC108311.1 | 15 | 2282 | AAI08312.1 | |
| BE675748.1 | 1 | 325 | ||
| BT007039.1 | AAP35688.1 | |||
| CR749690.1 | 2 | 1992 | CAH18474.2 | |
| R67284.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950700 | Chr.1:42731330 - 42756229 on Build GRCh38 | Deletion |
|
| esv3577249 | Chr.1:42576524 - 42885152 on Build GRCh38 | Gain |
|
| nsv829682 | Chr.1:42589429 - 42773568 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map