Assay Details
Target Gene Details
Entrez Gene ID: | 25852 |
Gene Name: | armadillo repeat containing 8 |
Gene Aliases: |
GID5, HSPC056, S863-2, VID28 |
Location: |
Chr.3:138187248-138298389 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARMC8 | NM_001267041.2 | 21 | 3681 | NP_001253970.1 |
| NM_001267042.2 | 21 | 3573 | NP_001253971.1 | |
| NM_001282342.1 | 20 | 3604 | NP_001269271.1 | |
| NM_015396.5 | 23 | 3850 | NP_056211.2 | |
| XM_006713565.2 | 23 | 3810 | XP_006713628.1 | |
| XM_011512632.1 | 22 | 3717 | XP_011510934.1 | |
| XM_011512633.1 | 22 | 3771 | XP_011510935.1 | |
| XM_017006095.1 | 22 | 3571 | XP_016861584.1 | |
| XM_017006097.1 | 21 | 3517 | XP_016861586.1 | |
| XM_017006098.1 | 20 | 3463 | XP_016861587.1 | |
| XM_017006099.1 | 21 | 3554 | XP_016861588.1 | |
| AL096748.1 | 9 | 2871 | CAB46423.1 | |
| BQ719381.1 |
Target Gene Details
Entrez Gene ID: | 347736 |
Gene Name: | NME/NM23 family member 9 |
Gene Aliases: |
NM23-H9, TXL-2, TXL2, TXNDC6 |
Location: |
Chr.3:138261437-138331785 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NME9 | NM_178130.3 | NP_835231.1 | ||
| XM_005247429.3 | 12 | 5332 | XP_005247486.1 | |
| XM_006713615.3 | 11 | 7214 | XP_006713678.1 | |
| XM_011512774.2 | 11 | 5218 | XP_011511076.1 | |
| XM_017006317.1 | 10 | 5267 | XP_016861806.1 | |
| XM_017006318.1 | 9 | 5595 | XP_016861807.1 | |
| AK094830.1 | BAC04430.1 | |||
| AY336746.1 | AAQ16116.1 | |||
| BC107054.2 | AAI07055.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428422 | Chr.3:137344347 - 138686784 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map