Assay Details
Target Gene Details
Entrez Gene ID: | 1958 |
Gene Name: | early growth response 1 |
Gene Aliases: |
AT225, G0S30, KROX-24, NGFI-A, TIS8, ZIF-268, ZNF225 |
Location: |
Chr.5:138465492-138469315 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EGR1 | NM_001964.2 | 2 | 2951 | NP_001955.1 |
| M24019.1 | 1 | 329 | ||
| M62829.1 | 2 | 2935 | AAA35815.1 | |
| M80583.1 | 2 | 2929 | ||
| X52541.1 | 2 | 2951 | CAA36777.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5814n100 | Chr.5:138466715 - 138481256 on Build GRCh38 | Loss |
|
| nsv5025 | Chr.5:138453658 - 138483063 on Build GRCh38 | Deletion |
|
| dgv5813n100 | Chr.5:138461691 - 138483227 on Build GRCh38 | Loss |
|
| nsv528851 | Chr.5:138446515 - 138469963 on Build GRCh38 | Loss |
|
| nsv1034030 | Chr.5:138448826 - 138480010 on Build GRCh38 | Loss |
|
| dgv10088n54 | Chr.5:138465844 - 138479962 on Build GRCh38 | Loss |
|
| nsv5024 | Chr.5:138439431 - 138469706 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map