Assay Details
Target Gene Details
Entrez Gene ID: | 25913 |
Gene Name: | protection of telomeres 1 |
Gene Aliases: |
CMM10, GLM9, HPOT1 |
Location: |
Chr.7:124822386-124929983 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POT1 | NM_001042594.1 | 8 | 1107 | NP_001036059.1 |
| NM_015450.2 | 9 | 1238 | NP_056265.2 | |
| NR_003102.1 | 9 | 1238 | ||
| NR_003103.1 | 9 | 1238 | ||
| NR_003104.1 | 9 | 1238 | ||
| XM_006715917.3 | 9 | 1228 | XP_006715980.1 | |
| XM_011516006.1 | 10 | 1350 | XP_011514308.1 | |
| XM_011516007.1 | 10 | 1349 | XP_011514309.1 | |
| XM_017011941.1 | 9 | 1236 | XP_016867430.1 | |
| XM_017011942.1 | 8 | 1098 | XP_016867431.1 | |
| AK001230.1 | 5 | 660 | BAA91568.1 | |
| AK001935.1 | 6 | 767 | BAA91988.1 | |
| AK022580.1 | 9 | 1075 | BAB14110.1 | |
| AK026234.1 | BAB15404.1 | |||
| AK302263.1 | 9 | 1393 | ||
| AY823520.1 | 5 | 648 | AAW22613.1 | |
| AY823521.1 | 5 | 648 | AAW22614.1 | |
| AY823522.1 | 5 | 648 | AAW22615.1 | |
| AY823523.1 | 4 | 517 | AAW22616.1 | |
| AY823524.1 | 5 | 648 | AAW22617.1 | |
| BC002923.2 | 6 | 694 | AAH02923.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv824297 | Chr.7:124858539 - 124858979 on Build GRCh38 | Gain |
|
| nsv608341 | Chr.7:124846926 - 124932234 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map