Assay Details
Target Gene Details
Entrez Gene ID: | 63891 |
Gene Name: | ring finger protein 123 |
Gene Aliases: |
FP1477, KPC1 |
Location: |
Chr.3:49688753-49721529 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF123 | NM_022064.4 | NP_071347.2 | ||
| NR_135218.1 | ||||
| XM_006713285.1 | XP_006713348.1 | |||
| XM_011533995.1 | XP_011532297.1 | |||
| XM_017007018.1 | XP_016862507.1 | |||
| AF370367.1 | AAQ15203.1 | |||
| AK026968.1 | BAB15607.1 | |||
| AK090632.1 | ||||
| AK096353.1 | ||||
| AK126883.1 | ||||
| AK308130.1 | ||||
| AK308607.1 | ||||
| AL832034.1 | ||||
| AL832138.1 | ||||
| AL832344.1 | ||||
| AY744152.1 | AAU93470.1 | |||
| BC041145.1 | AAH41145.1 | |||
| BC057392.1 | AAH57392.2 | |||
| BC088801.1 | AAH88801.1 | |||
| BC130632.1 | ||||
| CR627051.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508217 | Chr.3:49578013 - 49732463 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map