Assay Details
Target Gene Details
Entrez Gene ID: | 10529 |
Gene Name: | nebulette |
Gene Aliases: |
LASP2, LNEBL |
Location: |
Chr.10:20779973-21174187 on Build GRCh38 |
Assay Gene Location: | Within Exon 38 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEBL | NM_001173484.1 | 7 | 5478 | NP_001166955.1 |
| NM_006393.2 | 28 | 7755 | NP_006384.1 | |
| NM_213569.2 | 7 | 5480 | NP_998734.1 | |
| XM_005252342.4 | 27 | 7757 | XP_005252399.1 | |
| XM_005252343.4 | 26 | 7616 | XP_005252400.1 | |
| XM_005252344.4 | 27 | 7660 | XP_005252401.1 | |
| XM_011519291.2 | 30 | 7751 | XP_011517593.1 | |
| XM_017015467.1 | 30 | 7867 | XP_016870956.1 | |
| XM_017015468.1 | 31 | 7635 | XP_016870957.1 | |
| AB209311.1 | 7 | 5352 | BAD92548.1 | |
| BI522541.1 | 1 | 315 | ||
| Y16241.1 | 28 | 7755 | CAA76130.1 |
More Information
Set Membership: |
|

Genomic Map