Assay Details
Target Gene Details
Entrez Gene ID: | 103504730 |
Gene Name: | microRNA 9500 |
Gene Aliases: |
hsa-mir-9500 |
Location: |
Chr.2:218823090-218823154 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR9500 | NR_128707.1 |
Target Gene Details
Entrez Gene ID: | 53632 |
Gene Name: | protein kinase AMP-activated non-catalytic subunit gamma 3 |
Gene Aliases: |
AMPKG3 |
Location: |
Chr.2:218821853-218831803 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKAG3 | NM_017431.2 | NP_059127.2 | ||
| XM_017004343.1 | 13 | 2202 | XP_016859832.1 | |
| AF214519.1 | AAF73987.1 | |||
| AJ249977.1 | CAB65117.1 | |||
| AK300078.1 | ||||
| W94830.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528407 | Chr.2:218822874 - 218833281 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map