Assay Details
Target Gene Details
Entrez Gene ID: | 81565 |
Gene Name: | nudE neurodevelopment protein 1 like 1 |
Gene Aliases: |
EOPA, MITAP1, NDE1L1, NDE2, NUDEL |
Location: |
Chr.17:8435852-8472744 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDEL1 | NM_001025579.2 | NP_001020750.1 | ||
| NM_030808.4 | NP_110435.1 | |||
| XM_011524013.1 | XP_011522315.1 | |||
| AF217798.3 | AAF24516.3 | |||
| AK056014.1 | ||||
| AK299559.1 | ||||
| BP201698.1 | ||||
| DB176966.1 | ||||
| DN993952.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509651 | Chr.17:8395733 - 8444515 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map