Assay Details
Target Gene Details
Entrez Gene ID: | 8565 |
Gene Name: | tyrosyl-tRNA synthetase |
Gene Aliases: |
CMTDIC, TYRRS, YRS, YTS |
Location: |
Chr.1:32775237-32818032 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YARS | NM_003680.3 | 7 | 1543 | NP_003671.1 |
| XM_011542347.1 | 5 | 542 | XP_011540649.1 | |
| XM_011542348.1 | 6 | 610 | XP_011540650.1 | |
| XM_017002651.1 | 8 | 916 | XP_016858140.1 | |
| AK026535.1 | 8 | 836 | ||
| AK125213.1 | 7 | 1664 | ||
| AK223608.1 | 7 | 869 | BAD97328.1 | |
| BC001933.1 | 7 | 777 | AAH01933.1 | |
| BC004151.2 | 7 | 777 | AAH04151.1 | |
| BC016689.1 | 7 | 802 | AAH16689.1 | |
| KU178431.1 | 7 | 755 | ||
| KU178432.1 | ||||
| U40714.1 | 7 | 755 | AAB39406.1 | |
| U89436.1 | 7 | 755 | AAB88409.1 |
More Information
Set Membership: |
|

Genomic Map