Assay Details
Target Gene Details
Entrez Gene ID: | 64800 |
Gene Name: | EF-hand calcium binding domain 6 |
Gene Aliases: |
DJBP, HSCBCIP1, dJ185D5.1 |
Location: |
Chr.22:43528549-43812403 on Build GRCh38 |
Assay Gene Location: | Within Exon 38 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFCAB6 | NM_022785.3 | NP_073622.2 | ||
| NM_198856.2 | NP_942153.1 | |||
| XM_005261704.2 | 31 | 4593 | XP_005261761.1 | |
| XM_005261705.3 | 30 | 4507 | XP_005261762.1 | |
| XM_011530316.1 | 32 | 4880 | XP_011528618.1 | |
| XM_011530317.2 | 30 | 4440 | XP_011528619.1 | |
| XM_011530318.2 | 30 | 4536 | XP_011528620.1 | |
| XM_011530319.2 | 29 | 4228 | XP_011528621.1 | |
| XM_011530320.2 | XP_011528622.1 | |||
| XM_011530321.2 | 29 | 4223 | XP_011528623.1 | |
| XM_011530322.2 | 30 | 4351 | XP_011528624.1 | |
| XM_011530323.2 | 27 | 4009 | XP_011528625.1 | |
| XM_011530325.2 | 25 | 3754 | XP_011528627.1 | |
| XM_011530327.2 | 22 | 3501 | XP_011528629.1 | |
| XM_011530329.1 | 18 | 2973 | XP_011528631.1 | |
| XM_017028910.1 | 32 | 4692 | XP_016884399.1 | |
| XM_017028911.1 | XP_016884400.1 | |||
| XM_017028912.1 | 28 | 4351 | XP_016884401.1 | |
| AA778573.1 | ||||
| AB073862.1 | BAB71780.1 | |||
| AK027241.1 | BAB15703.1 | |||
| AK225596.1 | ||||
| AK225605.1 | ||||
| AK292411.1 | ||||
| AK292498.1 | ||||
| AL355841.1 | CAB91065.1 | |||
| AW139921.1 | ||||
| BC039315.1 | AAH39315.1 |
Target Gene Details
Entrez Gene ID: | 100874197 |
Gene Name: | EFCAB6 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.22:43516254-43536605 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFCAB6-AS1 | NR_046563.1 | |||
| AK090843.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1058659 | Chr.22:43522577 - 43700374 on Build GRCh38 | Loss |
|
| nsv589098 | Chr.22:43463583 - 43718462 on Build GRCh38 | Gain |
|
| nsv471205 | Chr.22:43467417 - 43718981 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map