Assay Details
Target Gene Details
Entrez Gene ID: | 5161 |
Gene Name: | pyruvate dehydrogenase (lipoamide) alpha 2 |
Gene Aliases: |
PDHAL |
Location: |
Chr.4:95840088-95841474 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDHA2 | NM_005390.4 | 1 | 475 | NP_005381.1 |
| AK313872.1 | 1 | 435 | ||
| BC030697.2 | 1 | 471 | ||
| BC066953.1 | 1 | 457 | ||
| BC094760.1 | 1 | 470 | AAH94760.1 | |
| BC119656.1 | 1 | 443 | ||
| BC119657.1 | 1 | 443 | ||
| BC127637.1 | 1 | 443 | ||
| BC127638.1 | 1 | 442 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv10537 | Chr.4:95836420 - 96035143 on Build GRCh38 | Loss |
|
| nsv594915 | Chr.4:95707502 - 96165467 on Build GRCh38 | Gain |
|
| nsv997304 | Chr.4:95654047 - 95940198 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map