Assay Details
Target Gene Details
Entrez Gene ID: | 10569 |
Gene Name: | SLU7 homolog, splicing factor |
Gene Aliases: |
9G8, hSlu7 |
Location: |
Chr.5:160401641-160419161 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLU7 | NM_006425.4 | NP_006416.3 | ||
| XM_005265793.3 | XP_005265850.1 | |||
| XM_011534418.2 | XP_011532720.1 | |||
| AF101074.1 | AAD13774.1 | |||
| AK000813.1 | ||||
| AK024875.1 | ||||
| AK314564.1 | ||||
| BC010634.1 | AAH10634.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527693 | Chr.5:160405901 - 160411656 on Build GRCh38 | Loss |
|
| nsv5097 | Chr.5:160371035 - 160413932 on Build GRCh38 | Deletion |
|
| esv3607367 | Chr.5:160136720 - 160414186 on Build GRCh38 | Gain |
|
| nsv1015634 | Chr.5:159750943 - 160620377 on Build GRCh38 | Gain |
|
| esv3607370 | Chr.5:160306540 - 160422741 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map