Assay Details
Target Gene Details
Entrez Gene ID: | 83697 |
Gene Name: | solute carrier family 4 member 9 |
Gene Aliases: |
AE4 |
Location: |
Chr.5:140359645-140375177 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509090 | Chr.5:140311306 - 140421024 on Build GRCh38 | Insertion |
|
| nsv1020011 | Chr.5:140340815 - 140396254 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map