Assay Details
Target Gene Details
Entrez Gene ID: | 1545 |
Gene Name: | cytochrome P450 family 1 subfamily B member 1 |
Gene Aliases: |
CP1B, CYPIB1, GLC3A, P4501B1 |
Location: |
Chr.2:38067603-38076181 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP1B1 | NM_000104.3 | 2 | 1374 | NP_000095.2 |
| AK297039.1 | 2 | 957 | ||
| AK303862.1 | ||||
| AK312686.1 | 2 | 1376 | ||
| BC012049.1 | 2 | 1327 | AAH12049.1 | |
| BT019979.1 | 1 | 971 | AAV38782.1 | |
| BT020001.1 | 1 | 971 | AAV38804.1 | |
| U03688.1 | 2 | 1317 | AAA19567.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2659660 | Chr.2:38005484 - 38133194 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map