Assay Details
Target Gene Details
Entrez Gene ID: | 79345 |
Gene Name: | olfactory receptor family 51 subfamily B member 2 (gene/pseudogene) |
Gene Aliases: |
HOR5'Beta3, OR51B1P |
Location: |
Chr.11:5323298-5324352 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR51B2 | NM_033180.4 | 1 | 337 | NP_149420.4 |
| BC117461.1 | 1 | 337 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046068 | Chr.11:5314713 - 6042512 on Build GRCh38 | Gain |
|
| nsv1048608 | Chr.11:5307286 - 5623949 on Build GRCh38 | Gain |
|
| nsv524400 | Chr.11:5307131 - 5395392 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map