Assay Details
Target Gene Details
Entrez Gene ID: | 359 |
Gene Name: | aquaporin 2 |
Gene Aliases: |
AQP-CD, WCH-CD |
Location: |
Chr.12:49950741-49958881 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AQP2 | NM_000486.5 | 4 | 1975 | NP_000477.1 |
| AK055824.1 | 1 | 50 | ||
| AK225940.1 | ||||
| BC042496.1 | 4 | 1970 | AAH42496.1 |
Target Gene Details
Entrez Gene ID: | 101927318 |
Gene Name: | uncharacterized LOC101927318 |
Gene Aliases: |
- |
Location: |
Chr.12:49951512-49962924 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927318 | NR_110590.1 | |||
| NR_110591.1 | ||||
| BM839521.1 | ||||
| DB200798.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832407 | Chr.12:49849485 - 50018453 on Build GRCh38 | Loss |
|
| nsv558837 | Chr.12:49955982 - 50073986 on Build GRCh38 | Loss |
|
| nsv522972 | Chr.12:49802611 - 50017048 on Build GRCh38 | Gain |
|
| nsv558835 | Chr.12:49881354 - 50039451 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map