Assay Details
Target Gene Details
Entrez Gene ID: | 54752 |
Gene Name: | fibronectin type III domain containing 8 |
Gene Aliases: |
- |
Location: |
Chr.17:35121612-35130732 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FNDC8 | NM_017559.2 | 3 | 756 | NP_060029.1 |
| AK225743.1 | 3 | 721 | ||
| AK313775.1 | 3 | 789 | ||
| AL133054.1 | 3 | 707 | CAB61380.1 | |
| BC024002.2 | 3 | 737 | AAH24002.1 |
Target Gene Details
Entrez Gene ID: | 54475 |
Gene Name: | notchless homolog 1 |
Gene Aliases: |
Nle |
Location: |
Chr.17:35128755-35142315 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLE1 | XM_017024777.1 | 13 | 4443 | XP_016880266.1 |
| AB209111.1 | 12 | 4496 | BAD92348.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2036 | Chr.17:35124777 - 35143140 on Build GRCh38 | Insertion |
|
| esv3892998 | Chr.17:35106468 - 35295755 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map