Assay Details
Target Gene Details
Entrez Gene ID: | 80221 |
Gene Name: | acyl-CoA synthetase family member 2 |
Gene Aliases: |
ACSMW, AVYV493 |
Location: |
Chr.17:50426158-50474845 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACSF2 | NM_001288968.1 | NP_001275897.1 | ||
| NM_001288969.1 | NP_001275898.1 | |||
| NM_001288970.1 | NP_001275899.1 | |||
| NM_001288971.1 | NP_001275900.1 | |||
| NM_001288972.1 | NP_001275901.1 | |||
| NM_025149.5 | NP_079425.3 | |||
| NR_110232.1 | ||||
| XM_006722110.3 | XP_006722173.1 | |||
| XM_011525294.1 | XP_011523596.1 | |||
| AK024573.1 | BAB14930.1 | |||
| AK225044.1 | ||||
| AK225561.1 | ||||
| AK294205.1 | ||||
| AK295258.1 | ||||
| AK300145.1 | ||||
| AK300625.1 | ||||
| AK308584.1 | ||||
| AK311109.1 | ||||
| AY358660.1 | AAQ89023.1 | |||
| BC012053.1 | AAH12053.1 | |||
| BC014123.2 | AAH14123.1 |
Target Gene Details
Entrez Gene ID: | 1101 |
Gene Name: | chondroadherin |
Gene Aliases: |
SLRR4A |
Location: |
Chr.17:50464492-50469881 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHAD | XM_011524214.2 | 1 | 923 | XP_011522516.1 |
| AK292177.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952340 | Chr.17:50459540 - 50478639 on Build GRCh38 | Deletion |
|
| nsv833476 | Chr.17:50316997 - 50473985 on Build GRCh38 | Loss |
|
| nsv2081 | Chr.17:50455308 - 50499820 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map