Assay Details
Target Gene Details
Entrez Gene ID: | 390201 |
Gene Name: | olfactory receptor family 10 subfamily V member 1 |
Gene Aliases: |
OR11-256 |
Location: |
Chr.11:59712916-59713845 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR10V1 | NM_001005324.1 | 1 | 108 | NP_001005324.1 |
More Information
Set Membership: |
|

Genomic Map