Assay Details
Target Gene Details
Entrez Gene ID: | 7041 |
Gene Name: | transforming growth factor beta 1 induced transcript 1 |
Gene Aliases: |
ARA55, HIC-5, HIC5, TSC-5 |
Location: |
Chr.16:31472155-31477960 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGFB1I1 | NM_001042454.2 | NP_001035919.1 | ||
| NM_001164719.1 | NP_001158191.1 | |||
| NM_015927.4 | NP_057011.2 | |||
| AB007836.1 | BAA24799.1 | |||
| AF116343.1 | AAD22552.1 | |||
| AK122975.1 | ||||
| AK313327.1 | ||||
| BC001507.3 | AAH01507.2 | |||
| BC001830.2 | AAH01830.1 | |||
| BC017288.2 | AAH17288.1 | |||
| BC032545.1 | AAH32545.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833194 | Chr.16:31346858 - 31517224 on Build GRCh38 | Loss |
|
| esv3570465 | Chr.16:31450067 - 31915765 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map