Assay Details
Target Gene Details
Entrez Gene ID: | 6659 |
Gene Name: | SRY-box 4 |
Gene Aliases: |
EVI16 |
Location: |
Chr.6:21593741-21598619 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOX4 | NM_003107.2 | 1 | 483 | NP_003098.1 |
| AF070669.1 | 1 | 473 | ||
| AF124147.1 | 1 | 456 | ||
| BC072668.1 | 1 | 384 | AAH72668.1 | |
| BM817788.1 | 1 | 483 | ||
| BM822945.1 | ||||
| X70683.1 | CAA50018.1 |
More Information
Set Membership: |
|

Genomic Map