Assay Details
Target Gene Details
Entrez Gene ID: | 6470 |
Gene Name: | serine hydroxymethyltransferase 1 |
Gene Aliases: |
CSHMT, SHMT |
Location: |
Chr.17:18327860-18363563 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHMT1 | NM_001281786.1 | 11 | 1829 | NP_001268715.1 |
| NM_004169.4 | 12 | 1945 | NP_004160.3 | |
| NM_148918.2 | 11 | 1828 | NP_683718.1 | |
| XM_005256767.3 | 13 | 2016 | XP_005256824.1 | |
| XM_011523992.2 | 10 | 1673 | XP_011522294.1 | |
| XM_017024957.1 | 12 | 1770 | XP_016880446.1 | |
| XM_017024958.1 | 12 | 1899 | XP_016880447.1 | |
| XM_017024959.1 | 9 | 1520 | XP_016880448.1 | |
| XM_017024960.1 | 8 | 1397 | XP_016880449.1 | |
| AK223552.1 | 12 | 1876 | BAD97272.1 | |
| AK298415.1 | ||||
| BC007979.2 | 12 | 1791 | AAH07979.1 | |
| BC038598.1 | 12 | 1802 | AAH38598.1 | |
| BM792235.1 | 1 | 72 | ||
| Y14486.1 | 7 | 2056 | CAB54839.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833386 | Chr.17:18266947 - 18429412 on Build GRCh38 | Gain |
|
| esv1004827 | Chr.17:18324454 - 18330561 on Build GRCh38 | Deletion |
|
| nsv428333 | Chr.17:18296767 - 18543070 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map