Assay Details
Target Gene Details
Entrez Gene ID: | 3859 |
Gene Name: | keratin 12 |
Gene Aliases: |
K12 |
Location: |
Chr.17:40861178-40867210 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT12 | NM_000223.3 | 8 | 1597 | NP_000214.1 |
| CV573989.1 | 2 | 241 | ||
| D78367.1 | 8 | 1597 | BAA11376.1 |
More Information
Set Membership: |
|

Genomic Map