Assay Details
Target Gene Details
Entrez Gene ID: | 84833 |
Gene Name: | up-regulated during skeletal muscle growth 5 homolog (mouse) |
Gene Aliases: |
DAPIT, HCVFTP2, bA792D24.4 |
Location: |
Chr.10:103389050-103396513 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USMG5 | NM_001206426.1 | NP_001193355.1 | ||
| NM_001206427.1 | NP_001193356.1 | |||
| NM_032747.3 | NP_116136.1 | |||
| AJ272056.1 | CAC81243.1 | |||
| AK311888.1 | ||||
| AL110185.1 | ||||
| BC007087.1 | AAH07087.1 | |||
| BC072683.1 | AAH72683.1 | |||
| BC091507.1 | AAH91507.1 | |||
| CN367000.1 | ||||
| DW446355.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv965n100 | Chr.10:103385173 - 103457152 on Build GRCh38 | Gain |
|
| nsv7529 | Chr.10:103394163 - 103424722 on Build GRCh38 | Insertion |
|
| dgv280e212 | Chr.10:103390805 - 103464349 on Build GRCh38 | Gain |
|
| esv3577694 | Chr.10:103391276 - 103553473 on Build GRCh38 | Gain |
|
| esv3624413 | Chr.10:103392636 - 103459758 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map