Assay Details
Target Gene Details
Entrez Gene ID: | 79639 |
Gene Name: | transmembrane protein 53 |
Gene Aliases: |
NET4 |
Location: |
Chr.1:44653247-44674608 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM53 | NM_001300746.1 | 2 | 1212 | NP_001287675.1 |
| NM_001300747.1 | 3 | 1244 | NP_001287676.1 | |
| NM_001300748.1 | 3 | 1241 | NP_001287677.1 | |
| NM_024587.3 | 3 | 1334 | NP_078863.2 | |
| XM_011542138.2 | 3 | 1463 | XP_011540440.1 | |
| AK026006.1 | 3 | 1141 | BAB15317.1 | |
| AK225405.1 | 3 | 1141 | ||
| AK295356.1 | 2 | 934 | ||
| AK310016.1 | 3 | 1278 | ||
| AL832539.1 | 2 | 1046 | CAI46125.1 | |
| BC064520.1 | 3 | 1153 | AAH64520.1 | |
| BM806701.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461395 | Chr.1:44433326 - 44654293 on Build GRCh38 | Gain |
|

Genomic Map