Assay Details
Target Gene Details
Entrez Gene ID: | 25807 |
Gene Name: | rhomboid domain containing 3 |
Gene Aliases: |
C22orf3, HS984G1A, PTAG |
Location: |
Chr.22:29259855-29268196 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHBDD3 | NM_012265.1 | 7 | 1472 | NP_036397.1 |
| XM_006724224.3 | 6 | 1225 | XP_006724287.1 | |
| XM_011530107.2 | 7 | 1402 | XP_011528409.1 | |
| XM_017028749.1 | 4 | 1988 | XP_016884238.1 | |
| XM_017028750.1 | 6 | 1790 | XP_016884239.1 | |
| AL050346.1 | 7 | 1472 | CAB43551.1 | |
| BC002705.2 | 5 | 1123 | AAH02705.1 | |
| CR456411.1 | 6 | 1178 | CAG30297.1 | |
| CR457382.1 | 5 | 1100 | CAG33663.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834171 | Chr.22:29230413 - 29433855 on Build GRCh38 | Loss |
|
| nsv829154 | Chr.22:29221496 - 29328012 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map