Assay Details
Target Gene Details
Entrez Gene ID: | 50617 |
Gene Name: | ATPase H+ transporting V0 subunit a4 |
Gene Aliases: |
A4, ATP6N1B, ATP6N2, RDRTA2, RTA1C, RTADR, STV1, VPH1, VPP2 |
Location: |
Chr.7:138706294-138799839 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP6V0A4 | NM_020632.2 | NP_065683.2 | ||
| NM_130840.2 | NP_570855.2 | |||
| XM_005250393.1 | XP_005250450.1 | |||
| XM_005250394.3 | XP_005250451.1 | |||
| AF245517.1 | AAG11415.1 | |||
| AK292945.1 | ||||
| BC109304.1 | AAI09305.1 | |||
| BC109305.1 | AAI09306.1 | |||
| DB230144.1 |
Target Gene Details
Entrez Gene ID: | 155006 |
Gene Name: | transmembrane protein 213 |
Gene Aliases: |
- |
Location: |
Chr.7:138797487-138806759 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM213 | NM_001085429.1 | NP_001078898.1 | ||
| XM_005250167.3 | 1 | 566 | XP_005250224.1 | |
| AK055841.1 | ||||
| BC131697.1 | 1 | 105 | ||
| DA626421.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831154 | Chr.7:138785743 - 138989367 on Build GRCh38 | Gain |
|
| esv2735212 | Chr.7:138060002 - 140415947 on Build GRCh38 | Deletion |
|
| esv3615199 | Chr.7:138793218 - 138823621 on Build GRCh38 | Gain |
|
| esv3615200 | Chr.7:138794105 - 138836379 on Build GRCh38 | Gain |
|
| nsv1030956 | Chr.7:138478424 - 138894773 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map