Assay Details
Target Gene Details
Entrez Gene ID: | 9986 |
Gene Name: | Ras converting CAAX endopeptidase 1 |
Gene Aliases: |
FACE2, RCE1A, RCE1B |
Location: |
Chr.11:66843412-66846532 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RCE1 | NM_001032279.1 | 5 | 571 | NP_001027450.1 |
| NM_005133.2 | 5 | 575 | NP_005124.1 | |
| AF121951.1 | 5 | 575 | AAD22632.1 | |
| AL713740.1 | 4 | 1270 | ||
| BC052622.1 | 5 | 532 | ||
| BC093726.1 | 5 | 571 | AAH93726.1 | |
| BC093728.1 | 5 | 570 | AAH93728.1 | |
| Y13835.2 | 5 | 544 | CAB46278.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv364 | Chr.11:66835722 - 66847867 on Build GRCh38 | Insertion |
|
| nsv555226 | Chr.11:66812101 - 67077891 on Build GRCh38 | Gain |
|
| nsv951020 | Chr.11:66843130 - 66869629 on Build GRCh38 | Deletion |
|
| nsv508638 | Chr.11:66774182 - 66876163 on Build GRCh38 | Deletion |
|
| nsv555225 | Chr.11:66700998 - 66866926 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map