Assay Details
Target Gene Details
Entrez Gene ID: | 11007 |
Gene Name: | coiled-coil domain containing 85B |
Gene Aliases: |
DIPA |
Location: |
Chr.11:65890404-65891635 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC85B | NM_006848.2 | 1 | 424 | NP_006839.2 |
| AI583181.1 | 1 | 66 | ||
| AK312109.1 | 1 | 184 | ||
| BC008796.2 | 1 | 189 | AAH08796.1 | |
| U63825.1 | 1 | 72 | AAB05928.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759834 | Chr.11:65792422 - 65954892 on Build GRCh38 | Gain |
|
| nsv832191 | Chr.11:65864858 - 66038649 on Build GRCh38 | Loss |
|
| nsv951019 | Chr.11:65874630 - 65899129 on Build GRCh38 | Deletion |
|
| nsv555215 | Chr.11:65814108 - 65909045 on Build GRCh38 | Loss |
|
| nsv1159905 | Chr.11:65836608 - 65922038 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map