Assay Details
Target Gene Details
Entrez Gene ID: | 4354 |
Gene Name: | membrane palmitoylated protein 1 |
Gene Aliases: |
AAG12, DXS552E, EMP55, MRG1, PEMP |
Location: |
Chr.X:154778684-154805527 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPP1 | NM_001166460.1 | NP_001159932.1 | ||
| NM_001166461.1 | NP_001159933.1 | |||
| NM_001166462.1 | NP_001159934.1 | |||
| NM_002436.3 | NP_002427.1 | |||
| XM_011531167.1 | XP_011529469.1 | |||
| XM_011531169.1 | XP_011529471.1 | |||
| AK223396.1 | BAD97116.1 | |||
| AK290246.1 | ||||
| AK303111.1 | ||||
| AK304538.1 | ||||
| AK310681.1 | ||||
| AK311541.1 | ||||
| AK312296.1 | ||||
| AK315957.1 | ||||
| AY423731.1 | AAS00494.1 | |||
| AY634686.1 | AAV35469.1 | |||
| BC002392.2 | AAH02392.1 | |||
| DA074686.1 | ||||
| M64925.1 | AAA60059.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss |
|
| nsv516158 | Chr.X:154790808 - 154831402 on Build GRCh38 | Gain |
|
| esv29972 | Chr.X:154781072 - 155699751 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map