Assay Details
Target Gene Details
Entrez Gene ID: | 79711 |
Gene Name: | importin 4 |
Gene Aliases: |
Imp4 |
Location: |
Chr.14:24180219-24188918 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IPO4 | NM_024658.3 | 30 | 3413 | NP_078934.3 |
| NR_051979.1 | 30 | 3433 | ||
| AB208897.1 | 30 | 3302 | BAD92134.1 | |
| AF411122.1 | AAL78660.1 | |||
| AK026991.1 | 23 | 2588 | BAB15616.1 | |
| AK027871.1 | 12 | 1381 | BAB55421.1 | |
| AK056527.1 | 30 | 3354 | ||
| AK056702.1 | 30 | 3260 | ||
| AK057044.1 | 29 | 3394 | ||
| AK057277.1 | 30 | 3317 | ||
| AK074743.1 | 15 | 1678 | BAC11174.1 | |
| AK094107.1 | 30 | 3261 | ||
| AK094897.1 | 29 | 3420 | ||
| AK095306.1 | 30 | 3261 | ||
| AK302097.1 | 24 | 2471 | ||
| AK314305.1 | ||||
| BC003690.1 | 21 | 2400 | AAH03690.2 | |
| BC110804.1 | 30 | 3315 | AAI10805.1 | |
| BC136759.1 | 30 | 3364 | ||
| BX248267.1 | 30 | 3295 | CAD62595.1 | |
| CR457367.1 | CAG33648.1 |
Target Gene Details
Entrez Gene ID: | 9985 |
Gene Name: | REC8 meiotic recombination protein |
Gene Aliases: |
HR21spB, REC8L1, Rec8p |
Location: |
Chr.14:24171853-24180408 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REC8 | XM_011537421.1 | XP_011535723.1 | ||
| XM_017021841.1 | XP_016877330.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522667 | Chr.14:24172317 - 24185384 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map