Assay Details
Target Gene Details
Entrez Gene ID: | 118427 |
Gene Name: | olfactomedin 3 |
Gene Aliases: |
NOE3, NOELIN3, OPTIMEDIN |
Location: |
Chr.1:101800028-101997234 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OLFM3 | NM_001288821.1 | 6 | 981 | NP_001275750.1 |
| NM_001288823.1 | 7 | 1293 | NP_001275752.1 | |
| NM_058170.3 | 6 | 1232 | NP_477518.2 | |
| NR_110210.1 | 8 | 2030 | ||
| NR_110211.1 | 7 | 1969 | ||
| NR_110212.1 | 7 | 1718 | ||
| XM_017000240.1 | 7 | 1136 | XP_016855729.1 | |
| AF397392.2 | 6 | 874 | AAK97473.1 | |
| AF397393.2 | 7 | 1611 | AAK97474.1 | |
| AF397394.1 | 6 | 903 | AAK97475.1 | |
| AF397395.1 | 7 | 1640 | AAK97476.1 | |
| AF397396.1 | 7 | 964 | AAK97477.1 | |
| AF397397.1 | 8 | 1701 | AAK97478.1 | |
| AK095551.1 | 4 | 458 | ||
| AK095724.1 | 6 | 1028 | ||
| AK300870.1 | 6 | 981 | ||
| AY358722.1 | 6 | 945 | AAQ89084.1 | |
| BC022531.1 | 6 | 990 | AAH22531.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3582471 | Chr.1:101578331 - 102371530 on Build GRCh38 | Gain |
|
| nsv2143 | Chr.1:101788623 - 101823251 on Build GRCh38 | Insertion |
|
| nsv522921 | Chr.1:101704739 - 102410836 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map