Assay Details
Target Gene Details
Entrez Gene ID: | 3881 |
Gene Name: | keratin 31 |
Gene Aliases: |
HA1, Ha-1, KRTHA1, hHa1 |
Location: |
Chr.17:41393724-41397592 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT31 | NM_002277.2 | NP_002268.2 | ||
| AK313419.1 | ||||
| BC114468.1 | AAI14469.1 | |||
| X86570.1 | CAA60378.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2052 | Chr.17:41377018 - 41402618 on Build GRCh38 | Insertion |
|
| nsv483014 | Chr.17:41339595 - 41514244 on Build GRCh38 | Loss |
|
| esv2758690 | Chr.17:41145123 - 41740591 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map