Assay Details
Target Gene Details
Entrez Gene ID: | 9844 |
Gene Name: | engulfment and cell motility 1 |
Gene Aliases: |
CED-12, CED12, ELMO-1 |
Location: |
Chr.7:36852906-37449409 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELMO1 | NM_001206480.2 | 16 | 1882 | NP_001193409.1 |
| NM_001206482.1 | 16 | 2052 | NP_001193411.1 | |
| NM_014800.10 | 16 | 1693 | NP_055615.8 | |
| XM_005249919.2 | 17 | 2246 | XP_005249976.1 | |
| XM_006715805.1 | 16 | 1538 | XP_006715868.1 | |
| XM_011515654.1 | 17 | 2931 | XP_011513956.1 | |
| XM_017012838.1 | 17 | 3116 | XP_016868327.1 | |
| XM_017012839.1 | 16 | 1480 | XP_016868328.1 | |
| AF398885.1 | 15 | 1344 | AAL14466.1 | |
| AK126565.1 | 4 | 733 | BAC86597.1 | |
| AK291784.1 | 16 | 2052 | ||
| AK296253.1 | 13 | 1213 | ||
| AK314349.1 | 16 | 1482 | ||
| AL136787.1 | 16 | 1594 | CAB66721.1 | |
| BC114341.2 | 16 | 1431 | AAI14342.1 |
Target Gene Details
Entrez Gene ID: | 100861514 |
Gene Name: | ELMO1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.7:36997796-37013630 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELMO1-AS1 | NR_104120.1 | |||
| DA058843.1 |
More Information
Set Membership: |
|

Genomic Map