Assay Details
Target Gene Details
Entrez Gene ID: | 57834 |
Gene Name: | cytochrome P450 family 4 subfamily F member 11 |
Gene Aliases: |
CYPIVF11 |
Location: |
Chr.19:15912370-15934866 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP4F11 | NM_001128932.1 | NP_001122404.1 | ||
| NM_021187.3 | 1 | 361 | NP_067010.3 | |
| AF236085.1 | AAG15889.1 | |||
| AK309292.1 | 1 | 69 | ||
| BC016853.1 | AAH16853.1 | |||
| DC345817.1 | 1 | 330 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv821195 | Chr.19:15930809 - 15934869 on Build GRCh38 | Deletion |
|
| nsv833766 | Chr.19:15759095 - 15956904 on Build GRCh38 | Loss |
|
| esv21733 | Chr.19:15926814 - 15934869 on Build GRCh38 | Gain+Loss |
|
| nsv1064684 | Chr.19:15311319 - 16062011 on Build GRCh38 | Gain |
|
| esv2718244 | Chr.19:15743561 - 15951200 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map