Assay Details
Target Gene Details
Entrez Gene ID: | 84419 |
Gene Name: | chromosome 15 open reading frame 48 |
Gene Aliases: |
FOAP-11, NMES1 |
Location: |
Chr.15:45430529-45433449 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C15orf48 | NM_032413.3 | 4 | 535 | NP_115789.1 |
| NM_197955.2 | 5 | 421 | NP_922946.1 | |
| AB026707.1 | 5 | 315 | BAB61021.1 | |
| AF228422.1 | 4 | 456 | AAK00708.1 | |
| AK026298.1 | 4 | 453 | ||
| BC021173.2 | 5 | 341 | AAH21173.1 | |
| BM312626.1 | 5 | 385 | ||
| CA865860.1 | ||||
| DB447060.1 |
Target Gene Details
Entrez Gene ID: | 100126311 |
Gene Name: | microRNA 147b |
Gene Aliases: |
MIRN147B, mir-147b |
Location: |
Chr.15:45433050-45433129 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR147B | NR_030599.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469721 | Chr.15:45316683 - 45495804 on Build GRCh38 | Gain |
|
| esv3892684 | Chr.15:45307172 - 46327637 on Build GRCh38 | Gain |
|
| esv3569553 | Chr.15:45366990 - 46080626 on Build GRCh38 | Gain |
|
| nsv1160292 | Chr.15:45351161 - 46175480 on Build GRCh38 | Duplication |
|
| nsv569315 | Chr.15:45153958 - 45480250 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map