Assay Details
Target Gene Details
Entrez Gene ID: | 7450 |
Gene Name: | von Willebrand factor |
Gene Aliases: |
F8VWF, VWD |
Location: |
Chr.12:5948874-6124675 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 39 - Intron 40 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VWF | NM_000552.4 | NP_000543.2 | ||
| AK297600.1 | ||||
| AK298097.1 | ||||
| AK301216.1 | ||||
| BQ898017.1 | ||||
| M10321.1 | AAB59512.1 | |||
| X04385.1 | CAA27972.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv446e212 | Chr.12:5977099 - 5986108 on Build GRCh38 | Loss |
|
| nsv557191 | Chr.12:5972658 - 6106468 on Build GRCh38 | Loss |
|
| nsv468988 | Chr.12:5413390 - 6141521 on Build GRCh38 | Gain |
|
| esv3580088 | Chr.12:5598006 - 6123173 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112046757] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map