Assay Details
Target Gene Details
Entrez Gene ID: | 9054 |
Gene Name: | NFS1 cysteine desulfurase |
Gene Aliases: |
HUSSY-08, IscS, NIFS |
Location: |
Chr.20:35668688-35699365 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFS1 | NM_001198989.1 | 12 | 1825 | NP_001185918.1 |
| NM_021100.4 | 13 | 1978 | NP_066923.3 | |
| NR_037570.2 | 14 | 2100 | ||
| AF097025.2 | 13 | 1964 | AAD09187.2 | |
| AJ010952.1 | 9 | 1486 | CAA09424.1 | |
| AK001265.1 | 13 | 1963 | ||
| AK001470.1 | 13 | 1974 | ||
| AK056242.1 | 13 | 2249 | ||
| AK091049.1 | 5 | 2520 | ||
| AK223239.1 | BAD96959.1 | |||
| BC065560.1 | 13 | 1950 | AAH65560.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526188 | Chr.20:35576541 - 36019079 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map